Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab™ UroFix Prefills
An ideal fixative for prostate biopsies.
$41.60 - $83.70
Specifications
Packaging Type | Vial |
---|---|
For Use With (Application) | Tissue Fixation |
Alert:
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
STLUF0507
|
StatLab™
UF0507 |
96 x 20 mL |
Case of 96 for $41.60
|
|
|||||
STLUF0460
|
StatLab™
UF0460 |
96 x 120 mL |
Case of 96 for $83.70
|
|
|||||
STLUF0230
|
StatLab™
UF0230 |
96 x 60 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
Description
- Preserves nuclear detail very well
- Penetrates tissue quicker, thereby shortening fixation times
Specifications
Vial | |
Tissue Fixation |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title