Learn More
StatLab™ UroFix Prefills
SDP
An ideal fixative for prostate biopsies.
Supplier:  StatLab™ UF0460
 View more versions of this product
Catalog No. STLUF0460
Explore available promotions
Add to Cart
Alert:
Harmful if swallowed, if in contact with skin, or if inhaled.
This item is not returnable. View return policy
Description
Description
  • Preserves nuclear detail very well
  • Penetrates tissue quicker, thereby shortening fixation times
Specifications
Specifications
Vial
96 x 120 mL
Tissue Fixation