Learn More
StatLab™ UroFix Prefills
SDP
An ideal fixative for prostate biopsies.
Supplier:  StatLab™ UF0230
 View more versions of this product
Catalog No. STLUF0230
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Alert:
Harmful if swallowed, if in contact with skin, or if inhaled.
This item is not returnable. View return policy
Description
Description
  • Preserves nuclear detail very well
  • Penetrates tissue quicker, thereby shortening fixation times
Specifications
Specifications
Vial
96 x 60 mL
Tissue Fixation
Product Suggestions
Product Suggestions