Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ TSG Series General Purpose Chromatography Refrigerators
Designed for your everyday laboratory needs
Specifications
Description | TSG Series general purpose chromatography refrigerator |
---|---|
Temperature Range | 2°C to 10°C |
Temperature (Setpoint) | +5°C |
Refrigerant | R290 |
Capacity (English) Shelf | 100 lb./full shelf; 50 lb./half shelf |
Catalog Number | Mfr. No. | Door Style | Cabinet Material | Interior | Capacity (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Door Style | Cabinet Material | Interior | Capacity (Metric) | Price | Quantity & Availability | ||
TSG30CPGA
|
Thermo Scientific™
TSG30CPGA |
Glass | Stainless Steel | Painted White Steel | 764 L |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
TSG30CSGA
|
Thermo Scientific™
TSG30CSGA |
Glass | Gray-painted | Stainless Steel | 764 L | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
TSG45CPLA
|
Thermo Scientific™
TSG45CPLA |
Sliding Glass | Stainless Steel | Painted White Steel | 1274 L | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
TSG45CSLA
|
Thermo Scientific™
TSG45CSLA |
Sliding Glass | Gray-painted | Stainless Steel | 1274 L |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
TSG49CPGA
|
Thermo Scientific™
TSG49CPGA |
Glass | Stainless Steel | Painted White Steel | 1387 L |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
TSG49CSGA
|
Thermo Scientific™
TSG49CSGA |
Glass | Gray-painted | Stainless Steel | 1387 L | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
TSG72CPGA
|
Thermo Scientific™
TSG72CPGA |
Glass | Stainless Steel | Painted White Steel | 2038 L |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
TSG72CSGA
|
Thermo Scientific™
TSG72CSGA |
Glass | Gray-painted | Stainless Steel | 2038 L |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
Description
Thermo Scientific™ TSG Series general-purpose chromatography refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2°to 10°C controlled environment.
- Hydrocarbon safety sensor ensures cargo and lab personnel are kept saf
- Featuring a wide range of cabinet sizes from 30 cu. ft. to 72 cu. ft.
- Whisper-quiet operation
- ENERGY STAR certified
- Temperature, door ajar*, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
- 2 years parts and labor, plus 3 year compressor warranty
Specifications
TSG Series general purpose chromatography refrigerator | |
+5°C | |
100 lb./full shelf; 50 lb./half shelf | |
cULus, ENERGYSTAR, Prop 65 | |
1 Rear access port and 2 side access ports | |
Alarm | |
NEMA 5-15 P | |
115 V 60 Hz | |
Energy efficient |
2°C to 10°C | |
R290 | |
45 kg/full shelf, 22.5 kg/half shelf | |
Digital Microprocessor | |
Adaptive Defrost | |
Key Lock | |
115 V | |
60 Hz |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title