Learn More
Thermo Scientific™ TSG Series General Purpose Chromatography Refrigerators
Designed for your everyday laboratory needs
Supplier: Thermo Scientific™ TSG49CPGA
Description
Thermo Scientific™ TSG Series general-purpose chromatography refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2°to 10°C controlled environment.
• Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
• Featuring a wide range of cabinet sizes from 30 cu. ft. to 72 cu. ft.
• Whisper-quiet operation
• ENERGY STAR certified
• Temperature, door ajar*, and power failure (with battery back-up) alarms
• Green, natural refrigerants
• Adaptive defrost
• Glass (double pane) or solid door configurations
• Choice of white painted interior and exterior, or stainless steel interior and gray exterior
• Digital controller
• Rear access port
• Remote alarm contacts
• Keyed door lock(s)
• LED interior lighting
• Low heat output
• 2 years parts and labor, plus 3 year compressor warranty
*All models except TSG45CPLA and TSG45CSLA come equipped with door ajar alarms
Specifications
TSG Series general purpose chromatography refrigerator | |
1387 L | |
+5°C | |
Glass | |
8 Full, 2 Half | |
45 kg/full shelf, 22.5 kg/half shelf | |
White Painted | |
cULus, ENERGYSTAR, Prop 65 | |
1 Rear access port and 2 side access ports | |
Adaptive Defrost | |
Key Lock | |
32 x 52.7 x 81 in.(813 x 1339 x 813 mm) | |
NEMA 5-15 P | |
115 V 60 Hz | |
Energy efficient | |
271 kg |
49 cu. ft. | |
2°C to 10°C | |
R290 | |
2 | |
100 lb./full shelf; 50 lb./half shelf | |
Stainless Steel | |
Painted White Steel | |
Digital Microprocessor | |
2 years parts and labor, plus 3-year compressor warranty | |
Alarm | |
28.5 x 49 x 61.75 in. (724 x 1245 x 1569 mm) | |
8.8 A | |
115 V | |
60 Hz | |
597 lb. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.