Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSG Series General Purpose Chromatography Refrigerators

Designed for your everyday laboratory needs

Supplier:  Thermo Scientific™ TSG45CSLA

Request Bulk Order

 View more versions of this product

Catalog No. TSG45CSLA

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Explore available promotions
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Description

Description

Thermo Scientific™ TSG Series general-purpose chromatography refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2°to 10°C controlled environment.

• Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
• Featuring a wide range of cabinet sizes from 30 cu. ft. to 72 cu. ft.
• Whisper-quiet operation
• ENERGY STAR certified
• Temperature, door ajar*, and power failure (with battery back-up) alarms
• Green, natural refrigerants
• Adaptive defrost
• Glass (double pane) or solid door configurations
• Choice of white painted interior and exterior, or stainless steel interior and gray exterior
• Digital controller
• Rear access port
• Remote alarm contacts
• Keyed door lock(s)
• LED interior lighting
• Low heat output
• 2 years parts and labor, plus 3 year compressor warranty
*All models except TSG45CPLA and TSG45CSLA come equipped with door ajar alarms

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSG Series general purpose chromatography refrigerator
1274 L
+5°C
Sliding Glass
8 Full, 2 Half
45 kg/full shelf, 22.5 kg/half shelf
Painted
cULus, ENERGYSTAR, Prop 65
1 Rear access port and 2 side access ports
Adaptive Defrost
Key Lock
32 x 52.7 x 81 in.(813 x 1339 x 813 mm)
NEMA 5-15 P
115 V 60 Hz
Energy efficient
255 kg
45 cu. ft.
2°C to 10°C
R290
2
100 lb./full shelf; 50 lb./half shelf
Gray-painted
Stainless Steel
Digital Microprocessor
2 years parts and labor, plus 3-year compressor warranty
Alarm
27 x 49 x 61.75 in. (686 x 1245 x 1569 mm)
5 A
115 V
60 Hz
561 lb.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.