Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ SMART Spectro™ Spectrophometer and SMART 3™ Colorimeter Reagent Systems
For water analysis in the field or in the lab
$52.00 - $116.00
Specifications
For Use With (Application) | Water testing and environmental studies |
---|---|
For Use With (Equipment) | SMART Spectro Spectrophometer and SMART 3 Colorimeter |
Catalog Number | Mfr. No. | Type | Range | Test Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Range | Test Method | Price | Quantity & Availability | |||
S72372
|
LaMotte™
364101SC |
Aluminum | 0 to 0.3ppm | Eriochrome Cyanine R |
Each for $116.00
|
|
|||
S72361
|
LaMotte™
3645SC |
Chromium, Hexavalent | 0 to 1ppm | Diphenylcarbohydrazide |
Each for $52.00
|
|
|||
S72359
|
LaMotte™
3642SC |
Ammonia Nitrogen | 0 to 4ppm | Nesslerization |
Each for $66.75
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S72360
|
LaMotte™
3643SC |
Bromine, Low Range | 0 to 9ppm | DPD Tablets |
Each for $84.00
|
|
|||
S72364
|
LaMotte™
364901SC |
Nitrate Nitrogen | 0 to 3ppm | Cadmium Reduction |
Each for $73.50
|
|
|||
Description
- High-quality reagents manufactured from ACS raw materials for high reproducibility
- All systems are designed to overcome test interferences for fast and dependable results
Specifications
Water testing and environmental studies | |
SMART Spectro Spectrophometer and SMART 3 Colorimeter |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title