Learn More
LaMotte™ SMART Spectro™ Spectrophometer and SMART 3™ Colorimeter Reagent Systems
SCI_ED
For water analysis in the field or in the lab
Supplier:  LaMotte™ 3643SC
 View more versions of this product
Catalog No. S72360
Add to Cart
Description
Description
  • High-quality reagents manufactured from ACS raw materials for high reproducibility
  • All systems are designed to overcome test interferences for fast and dependable results
Specifications
Specifications
Water testing and environmental studies
100
DPD Tablets
SMART Spectro Spectrophometer and SMART 3 Colorimeter
0 to 9ppm
Bromine, Low Range
Product Suggestions
Product Suggestions