Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ High-capacity and ECD Vent Traps
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Includes connecting lines and mounting kit.
$132.00 - $344.00
Specifications
Connection Type | 0.12 in. Fitting |
---|---|
Includes | Connecting lines and mounting kit |
Length (English) | 6 in. |
Length (Metric) | 15.2 cm |
Diameter (English) Outer | 1 in. |
Catalog Number | Mfr. No. | Type | Packaging Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Packaging Quantity | Price | Quantity & Availability | ||||
06-710-915
![]() |
Restek™
20698 |
High Capacity Split Vent Trap | 1 Each |
Each for $155.00
|
|
||||
06-711-822
![]() |
Restek™
22017 |
ECD Vent Trap | 1 Each |
Each for $132.00
|
|
||||
06-717-795
![]() |
Restek™
20699 |
High Capacity Split Vent Trap | 5/Pk. |
Pack of 5 for $344.00
|
|
||||
Description
- High-Capacity Split Vent Trap reduces the release of hazardous materials from the capillary split vent into the lab
- ECD Vent Trap reduces the release of hazardous materials from the ECD vent into the lab
- Fittings: 1/8 in.
- Dimensions: 15.2 x 2.5cm (6 x 1 in.)
Specifications
0.12 in. Fitting | |
6 in. | |
1 in. |
Connecting lines and mounting kit | |
15.2 cm | |
2.5 cm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title