Learn More
Ahlstrom-Munksjo Qualitative Filter Paper
Ahlstrom-Munksjo qualitative filter paper is recommended for use in analytical methods, which determine or identify particulate constituents of a mixture irrespective of the amount present. They are often used in routine separation work that still requires high purity and consistent performance.
$33.00 - $939.00
Specifications
Format Disc
Type Qualitative Filter Paper
View More Specs
Products 25
Catalog Number Mfr. No. Diameter (Metric) Filtration Speed Description Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Filtration Speed Description Price Quantity & Availability  
12-007-196
View Documents
Ahlstrom-Munksjo
05400470
4.7 cm Fast Speed Low Ash
Pack for $38.25
Add to Cart
12-007-198
View Documents
Ahlstrom-Munksjo
05400700
7.0 cm Fast Speed Low Ash
Pack for $45.25
Add to Cart
12-007-200
View Documents
Ahlstrom-Munksjo
05401100
11.0 cm Fast Speed Low Ash
Pack for $71.50
Add to Cart
12-007-201
View Documents
Ahlstrom-Munksjo
05401250
12.5 cm Fast Speed Low Ash
Pack for $83.00
Add to Cart
12-007-203
View Documents
Ahlstrom-Munksjo
05401850
18.5 cm Fast Speed Low Ash
Pack for $143.00
Add to Cart
12-007-207
View Documents
Ahlstrom-Munksjo
05404000
40.0 cm Fast Speed Low Ash
Pack for $406.00
Add to Cart
12-007-220
View Documents
Ahlstrom-Munksjo
05502400
24.0 cm Fast Speed Hardened Ashless
Pack for $258.00
Add to Cart
12-007-226
View Documents
Ahlstrom-Munksjo
07400550
5.5 cm Medium Speed Ashless
Pack for $39.50
Add to Cart
12-007-228
View Documents
Ahlstrom-Munksjo
07400900
9.0 cm Medium Speed Ashless
Pack for $60.75
Add to Cart
12-007-235
View Documents
Ahlstrom-Munksjo
07403200
32.0 cm Medium Speed Ashless
Pack for $344.00
Add to Cart
12-007-237
View Documents
Ahlstrom-Munksjo
07500210
2.1 cm Medium Speed Hardened Ashless
Pack for $40.50
Add to Cart
12-007-239
View Documents
Ahlstrom-Munksjo
07500425
4.25 cm Medium Speed Hardened Ashless
Pack for $42.50
Add to Cart
12-007-241
View Documents
Ahlstrom-Munksjo
07500700
7.0 cm Medium Speed Hardened Ashless
Pack for $53.00
Add to Cart
12-007-247
View Documents
Ahlstrom-Munksjo
07502400
24.0 cm Medium Speed Hardened Ashless
Pack for $266.00
Add to Cart
12-007-249
View Documents
Ahlstrom-Munksjo
07503200
32.0 cm Medium Speed Hardened Ashless
Pack for $460.00
Add to Cart
12-007-251
View Documents
Ahlstrom-Munksjo
07505000
50.0 cm Medium Speed Hardened Ashless
Pack for $939.00
Add to Cart
12-007-266
View Documents
Ahlstrom-Munksjo
09500425
4.25 cm Slow Speed Hardened Ashless
Pack for $42.50
Add to Cart
12-007-273
View Documents
Ahlstrom-Munksjo
09501850
18.5 cm Slow Speed Hardened Ashless
Pack for $178.00
Add to Cart
12-007-275
View Documents
Ahlstrom-Munksjo
09502700
27.0 cm Slow Speed Hardened Ashless
Pack for $325.00
Add to Cart
12-007-295
View Documents
Ahlstrom-Munksjo
39100700
7.0 cm Slow Speed Low Ash
Pack for $33.00
Add to Cart
12-007-297
View Documents
Ahlstrom-Munksjo
39101100
11.0 cm Slow Speed Low Ash
Pack for $53.75
Add to Cart
12-007-309
View Documents
Ahlstrom-Munksjo
39200900
9.0 cm Medium Speed Low Ash
Pack for $44.75
Add to Cart
12-007-311
View Documents
Ahlstrom-Munksjo
39201250
12.5 cm Medium Speed Low Ash
Pack for $65.50
Add to Cart
12-007-313
View Documents
Ahlstrom-Munksjo
39201850
18.5 cm Medium Speed Low Ash
Pack for $124.00
Add to Cart
12-007-315
View Documents
Ahlstrom-Munksjo
39202700
27.0 cm Medium Speed Low Ash
Pack for $258.00
Add to Cart
Description
Description
  • For use with liquid clarification, qualitative analytical separations for precipitates, buffers filtration, soil analysis, food and beverage testing
  • High purity filters
  • Consistent performance
  • Available in a variety of sizes
Specifications
Specifications
Disc
Qualitative Filter Paper