Learn More
Ahlstrom-Munksjo Qualitative Filter Paper
Ahlstrom-Munksjo qualitative filter paper is recommended for use in analytical methods, which determine or identify particulate constituents of a mixture irrespective of the amount present. They are often used in routine separation work that still requires high purity and consistent performance.
Supplier: Ahlstrom-Munksjo 39201850
Description
- For use with liquid clarification, qualitative analytical separations for precipitates, buffers filtration, soil analysis, food and beverage testing
- High purity filters
- Consistent performance
- Available in a variety of sizes
Specifications
Low Ash | |
Medium Speed | |
18.5 cm |
Disc | |
Qualitative Filter Paper |