Learn More
Ahlstrom-Munksjo Qualitative Filter Paper
Ahlstrom-Munksjo qualitative filter paper is recommended for use in analytical methods, which determine or identify particulate constituents of a mixture irrespective of the amount present. They are often used in routine separation work that still requires high purity and consistent performance.
Supplier:  Ahlstrom-Munksjo 05404000
 View more versions of this product
Catalog No. 12-007-207
Add to Cart
Description
Description
  • For use with liquid clarification, qualitative analytical separations for precipitates, buffers filtration, soil analysis, food and beverage testing
  • High purity filters
  • Consistent performance
  • Available in a variety of sizes
Specifications
Specifications
Low Ash
Fast Speed
40.0 cm
Disc
Qualitative Filter Paper
Product Suggestions
Product Suggestions