Learn More
Eisco™ Plastic Filter Funnels
SCI_ED
Eisco™ Plastic Filter Funnels are constructed of polyethylene which is chemical, acid, and alkaline resistant.
$.45 - $9.75
Specifications
Material Polyethylene
Color Clear
View More Specs
Products 9
Catalog Number Mfr. No. Diameter (Metric)Top Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric)Top Height (Metric) Price Quantity & Availability  
S89473
SCI_ED
View Documents
Eisco™
CH0454A
50 mm 9 cm
Each for $0.50
Add to cart
S23817
SCI_ED
View Documents
Eisco™
CH0454A10
50 mm 9 cm
Pack of 10 for $4.70
Add to cart
S89474
SCI_ED
View Documents
Eisco™
CH0454B
62 mm 12 cm
Each for $0.50
Edge
Add to cart
S23818
SCI_ED
View Documents
Eisco™
CH0454B10
62 mm 12 cm
Pack of 10 for $1.26
Save 71%
Reg : $4.40
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Add to cart
S89475
SCI_ED
View Documents
Eisco™
CH0454C
75 mm 14 cm
Each for $0.45
Add to cart
S23819
SCI_ED
View Documents
Eisco™
CH0454C10
75 mm 14 cm
Pack of 10 for $5.35
Add to cart
S23821
SCI_ED
View Documents
Eisco™
CH0454D10
100 mm 15.2 cm
Pack of 10 for $9.75
Add to cart
S23820
SCI_ED
View Documents
Eisco™
CH0454D
100 mm 15.2 cm
Each for $1.25
Add to cart
S89476
SCI_ED
View Documents
Eisco™
CH0454E
150 mm 15.2 cm
Each for $4.10
Add to cart
Description
Description
  • Chemically resistant polyethylene funnel
  • Translucent
  • Durable for student use
  • Easy to clean
  • Autoclavable
Warranty and Services
1 year limited manufacturer warranty
Specifications
Specifications
Polyethylene
Clear