Learn More
Eisco™ Plastic Filter Funnels
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Eisco™ Plastic Filter Funnels are constructed of polyethylene which is chemical, acid, and alkaline resistant.
Supplier: Eisco™ CH0454C10
Description
- Chemically resistant polyethylene funnel
- Translucent
- Durable for student use
- Easy to clean
- Autoclavable
Warranty and Services
1 year limited manufacturer warranty
Specifications
75 mm | |
Polyethylene | |
Clear |
7.5cm | |
14 cm | |
10 Pk. |