Learn More
Thermo Scientific™ Phusion High-Fidelity PCR Master Mix with HF Buffer
The Thermo Scientific™ Phusion™ High-Fidelity PCR Master Mixes are high performance enzymes now available in convenient master mix format.
$264.00 - $1045.00
Specifications
Format | Tube |
---|---|
GC-Rich PCR Performance | Medium |
Polymerase | Phusion High-Fidelity DNA Polymerase |
Reaction Speed | Fast |
Product Line | Phusion |
Description
The Thermo Scientific™ Phusion™ High-Fidelity PCR Master Mixes are high performance enzymes now available in convenient master mix format.
Phusion High-Fidelity PCR Master Mix with HF Buffer and Phusion High-Fidelity PCR Master Mix with GC Buffer are convenient 2x mixes containing Phusion DNA Polymerase, nucleotides and optimized HF or GC reaction buffer including MgCl2. Only template and primers need to be added by the user.- High fidelity (error rate 4.4 x 10-7 in Phusion HF Buffer)
- Speed of the polymerase allows short extension times (15-30 s/kb)
- Robust reactions, minimal optimization needed
- Increased product yields with minimal enzyme amounts
Includes:
2x Phusion High-Fidelity PCR Master Mix with HF Buffer contains Phusion DNA Polymerase, 2x Phusion HF Buffer and 400μM of each dNTP. The master mix supplies 1.5 mM MgCl2 and 200μM dNTP in final reaction concentration. A separate tube of DMSO is provided with the product. 2x Phusion High-Fidelity PCR Master Mix with GC Buffer contains Phusion DNA Polymerase, 2x Phusion GC Buffer and 400μM of each dNTP. The master mix supplies 1.5 mM MgCl2 and 200μM dNTP in final reaction concentration. A separate tube of DMSO is provided with the product.
Recommended for:
Fast PCR, high-fidelity PCR, cloning, long or difficult amplicons, DHPLC, microarray
Specifications
Tube | |
Phusion High-Fidelity DNA Polymerase | |
Phusion | |
DHPLC | |
2X | |
No | |
SuperMix or Master Mix | |
DNA |
Medium | |
Fast | |
High-Fidelity PCR Master Mix | |
Cloning | |
52 X | |
Blunt | |
≤20 kb |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser≈s own internal research. No other patent rights are conveyed expressly, by implication, or by estoppels. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.