Learn More
DWK Life Sciences Kimble™ Kontes™ Wide Mouth Heavy Wall Class A Volumetric Flasks
Heavy wall design increases flask durability.
$191.10 - $320.50
Specifications
Material Borosilicate Glass
Neck Style Bulb, wide mouth
View More Specs
Includes: Pennyhead stopper
Products 7
Catalog Number Mfr. No. Length (Metric) Tolerance Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Tolerance Capacity (Metric) Price Quantity & Availability  
K623020-0100
View Documents Promotion Details
DWK Life Sciences
6230200100
200 mm ±0.08 mL 100 mL
Case for $191.10
Add to Cart
K623020-0200
View Documents Promotion Details
DWK Life Sciences
6230200200
230 mm ±0.12 mL 200 mL N/A N/A
This item has been discontinued and is no longer available. Please reach out to your sales representative.
K623020-1000
View Documents Promotion Details
DWK Life Sciences
6230201000
345 mm ±0.3 mL 1000 mL
Case for $320.50
Add to Cart
K623020-0400
View Documents Promotion Details
DWK Life Sciences
6230200400
290 mm ±0.15 mL 400 mL
Case for $269.00
Add to Cart
K623020-0300
View Documents Promotion Details
DWK Life Sciences
6230200300
250 mm ±0.12 mL 300 mL
Case for $239.50
Add to Cart
K623020-0250
View Documents
DWK Life Sciences
6230200250
230 mm ±0.12 mL 250 mL Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. View substitute products »
K623020-0500
View Documents Promotion Details
DWK Life Sciences
6230200500
305 mm ±0.15 mL 500 mL Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. View substitute products »
Description
Description
Heavy wall design increases flask durability
  • Wide mouth mixing bulb provides easy access into the flask for dissolving tablets and other solids
  • Mixing bulb in neck accommodates approximately one tenth of the flask's total volume
Specifications
Specifications
Borosilicate Glass
Bulb, wide mouth