Learn More
DWK Life Sciences Kimble™ Kontes™ Wide Mouth Heavy Wall Class A Volumetric Flasks
SDP
Heavy wall design increases flask durability.
Supplier:  DWK Life Sciences 6230200500
 View more versions of this product
Catalog No. K623020-0500
Add to Cart
Includes: Pennyhead stopper
Description
Description
Heavy wall design increases flask durability
  • Wide mouth mixing bulb provides easy access into the flask for dissolving tablets and other solids
  • Mixing bulb in neck accommodates approximately one tenth of the flask's total volume
Specifications
Specifications
500 mL
12 in.
305 mm
±0.15 mL
Borosilicate Glass
30.5 cm
Bulb, wide mouth
Product Suggestions
Product Suggestions