Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Wide Mouth Heavy Wall Class A Volumetric Flasks
SDP

Heavy wall design increases flask durability.

Supplier:  DWK Life Sciences 6230200250

 View more versions of this product

Catalog No. K623020-0250


Add to Cart

Includes: Pennyhead stopper

Description

Description

Heavy wall design increases flask durability

  • Wide mouth mixing bulb provides easy access into the flask for dissolving tablets and other solids
  • Mixing bulb in neck accommodates approximately one tenth of the flask's total volume

Specifications

Specifications

250 mL
9.05 in.
230 mm
Case of 1
16
Borosilicate Glass
23 cm
Bulb, wide mouth
±0.12 mL
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit