Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSC Go Science Crazy Graham Coil Condensers
Sealed inner and outer tubes prevent leakage.
$44.00 - $44.25
Specifications
Material | Borosilicate Glass |
---|---|
Weight (English) Shipping | 1 lb. |
Weight (Metric) Shipping | 0.45 kg |
Description
- Made of borosilicate glass
- Inner coil provides additional surface area for coiling
- Ground glass joints 24/40 allows precision connection to jointed glassware
- Choose sizes from 250mm to 600mm
Specifications
Borosilicate Glass | |
1 lb. | |
0.45 kg |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title