Learn More
GSC Go Science Crazy Graham Coil Condensers
Sealed inner and outer tubes prevent leakage.
Supplier: GSC Go Science Crazy 4005400
Description
- Made of borosilicate glass
- Inner coil provides additional surface area for coiling
- Ground glass joints 24/40 allows precision connection to jointed glassware
- Choose sizes from 250mm to 600mm
Specifications
Borosilicate Glass | |
400 mm | |
1 lb. | |
0.45 kg |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov