Learn More
GSC Go Science Crazy Graham Coil Condensers
SCI_ED
Sealed inner and outer tubes prevent leakage.
Supplier:  GSC Go Science Crazy 4005300
 View more versions of this product
Catalog No. S04819A
Add to Cart
Description
Description
  • Made of borosilicate glass
  • Inner coil provides additional surface area for coiling
  • Ground glass joints 24/40 allows precision connection to jointed glassware
  • Choose sizes from 250mm to 600mm
Specifications
Specifications
Borosilicate Glass
1 lb.
300 mm
0.45 kg
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov