Learn More
National Public Seating™ Designer Science Table
High end designer adjustable legs for science and art environments
$1179.00 - $2076.00
Specifications
Certifications/Compliance | MAS2 |
---|---|
For Use With (Application) | Art and Science Environments |
Load Bearing Capacity (English) | 500 lb. |
Style | Black legs |
Warranty | 10 years |
Catalog Number | Mfr. No. | Dimensions (D x W x H) | Top Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (D x W x H) | Top Style | Price | Quantity & Availability | ||||
S83731
|
National Public Seating™
SLT62448B |
48 x 24 x 27 to 42 in. | Butcherblock |
Each for $1,584.00
|
|
||||
S83716
|
National Public Seating™
SLT62448C |
48 x 24 x 27 to 42 in. | Chemical Resistant |
Each for $1,179.00
|
|
||||
S83711
|
National Public Seating™
SLT62448H |
48 x 24 x 27 to 42 in. | HPL |
Each for $1,180.00
|
|
||||
S83721
|
National Public Seating™
SLT62448P |
48 x 24 x 27 to 42 in. | Phenolic |
Each for $1,349.00
|
|
||||
S83732
|
National Public Seating™
SLT62454B |
54 x 24 x 27 to 42 in. | Butcherblock |
Each for $2,076.00
|
|
||||
S83717
|
National Public Seating™
SLT62454C |
54 x 24 x 27 to 42 in. | Chemical Resistant |
Each for $1,256.00
|
|
||||
S83712
|
National Public Seating™
SLT62454H |
54 x 24 x 27 to 42 in. | HPL |
Each for $1,259.00
|
|
||||
S83722
|
National Public Seating™
SLT62454P |
54 x 24 x 27 to 42 in. | Phenolic |
Each for $1,499.00
|
|
||||
S83733
|
National Public Seating™
SLT62460B |
60 x 24 x 27 to 42 in. | Butcherblock |
Each for $1,796.00
|
|
||||
S83718
|
National Public Seating™
SLT62460C |
60 x 24 x 27 to 42 in. | Chemical Resistant |
Each for $1,270.00
|
|
||||
S83713
|
National Public Seating™
SLT62460H |
60 x 24 x 27 to 42 in. | HPL |
Each for $1,272.00
|
|
||||
S83723
|
National Public Seating™
SLT62460P |
60 x 24 x 27 to 42 in. | Phenolic |
Each for $1,542.00
|
|
||||
S83734
|
National Public Seating™
SLT62472B |
72 x 24 x 27 to 42 in. | Butcherblock |
Each for $1,940.00
|
|
||||
S83719
|
National Public Seating™
SLT62472C |
72 x 24 x 27 to 42 in. | Chemical Resistant |
Each for $1,333.00
|
|
||||
S83714
|
National Public Seating™
SLT62472H |
72 x 24 x 27 to 42 in. | HPL |
Each for $1,336.00
|
|
||||
S83724
|
National Public Seating™
SLT62472P |
72 x 24 x 27 to 42 in. | Phenolic |
Each for $1,566.00
|
|
||||
S83735
|
National Public Seating™
SLT63060B |
60 x 30 x 27 to 42 in. | Butcherblock |
Each for $1,974.00
|
|
||||
S83720
|
National Public Seating™
SLT63060C |
60 x 30 x 27 to 42 in. | Chemical Resistant |
Each for $1,370.00
|
|
||||
S83715
|
National Public Seating™
SLT63060H |
60 x 30 x 27 to 42 in. | HPL |
Each for $1,376.00
|
|
||||
S83725
|
National Public Seating™
SLT63060P |
60 x 30 x 27 to 42 in. | Phenolic |
Each for $1,650.00
|
|
||||
Description
- Under table hat support for 500 lb. weight capacity
- Tables adjust in 3/4 in. increments from 27 to 42 in. Height
- 11-gauge leg-bases are fastened to the tabletop with 6 heavy-duty screws
- All legs come standard with simple-adjust glides
- 10 Year Warranty
Specifications
MAS2 | |
500 lb. | |
10 years |
Art and Science Environments | |
Black legs | |
Table |