Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Public Seating™ Designer Science Table
SCI_ED

High end designer adjustable legs for science and art environments

Supplier:  National Public Seating™ SLT62472P

 View more versions of this product

Catalog No. S83724


Add to Cart

Description

Description

  • Under table hat support for 500 lb. weight capacity
  • Tables adjust in 3/4 in. increments from 27 to 42 in. Height
  • 11-gauge leg-bases are fastened to the tabletop with 6 heavy-duty screws
  • All legs come standard with simple-adjust glides
  • 10 Year Warranty
Specifications

Specifications

Table
Black
500 lb.
10 years
Art and Science Environments
Phenolic
MAS2
Black legs
72 x 24 x 27 to 42 in.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit