Learn More
SP Scienceware™ Bel-Art™ Sterileware Double Bagged Sterile Scoop Sampling System
Bagged Twice for Extra Protection from Contamination
Supplier:  SP Scienceware™ H369221010
 View more versions of this product
Catalog No. 03-411-817
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
Double bagging reduces contamination in multi-zone clean rooms while the included lid eliminates the cost of separate sterile bags or containers.
  • Ten individually packaged scoops are sealed in an outer bag to create multiples layers of protection against contaminants and debris
  • Snap-on lid allows scoop to be used as a container for transport; lock-tab can be bent under for a tighter seal
  • Sampling data can be marked on the lid with wax pencil, pre-marked label or labeling tape to facilitate tracking
  • Scoops remain upright when placed on a flat surface; can be used as weighing boats
  • Long handle increases the distance between the hand and sample to reduce contamination potential
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10-6
  • Lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
Specifications
Specifications
Scoop
60 mL
Certificate of Processing for Sterilization
20.5 x 4.1 x 2.9 cm (8 1/16 x 1 5/8 x 1 1/8 in.)
12.7 cm
2 oz.
Sterile
10/Pk.
5 in.
Product Suggestions
Product Suggestions