Assay Details
Amplicon Length: | 89 |
Chromosome Location: | Chr. 19 - 47227196 - 47227196 [+] on 47227196 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3190-3p | MIMAT0022839 |
Human | hsa-miR-3190-5p | MIMAT0015073 |
Human | hsa-miR-3191-5p | MIMAT0022732 |
Human | hsa-miR-3191-3p | MIMAT0015075 |
Stem-loop Details
Human
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)