Assay Details
Gene Family ID: | MIPF0001251, mir-3190 |
Mature miRNA Sequence: | UGUGGAAGGUAGACGGCCAGAGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3190-3p | MIMAT0022839 |
Stem-loop Details
Human
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3190-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0014235 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.19: 47226942 - 47227021 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UGUGGAAGGUAGACGGCCAGAGA | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0001251, mir-3190 |
Mature miRNA Sequence: | UGUGGAAGGUAGACGGCCAGAGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3190-3p | MIMAT0022839 |
Stem-loop ID | hsa-mir-3190 |
Stem-loop Accession # | MI0014235 |
Stem-loop Sequence | CUGGGGUCACCUGUCUGGCCAGCUACGUCCCCACGGCCCUUGUCAGUGUGGAAGGUAGACGGCCAGAGAGGUGACCCCGG |
Chromosome Location | Chr. 19 - 47226942 - 47227021 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0022839
miRBase ID: hsa-miR-3190-3p Mature miRNA Sequence: UGUGGAAGGUAGACGGCCAGAGA Chromosome Location: Chr. 19 - 47226942 - 47227021 [+] on Build GRCh38 |