Assay Details
Gene Family ID: | MIPF0001251, mir-3190 |
Mature miRNA Sequence: | CUCUCUGGCCGUCUACCUUCCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3191-5p | MIMAT0022732 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3191-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0014236 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.19: 47226944 - 47227019 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | CUCUCUGGCCGUCUACCUUCCA | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0001251, mir-3190 |
Mature miRNA Sequence: | CUCUCUGGCCGUCUACCUUCCA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3191-5p | MIMAT0022732 |
Stem-loop ID | hsa-mir-3191 |
Stem-loop Accession # | MI0014236 |
Stem-loop Sequence | GGGGUCACCUCUCUGGCCGUCUACCUUCCACACUGACAAGGGCCGUGGGGACGUAGCUGGCCAGACAGGUGACCCC |
Chromosome Location | Chr. 19 - 47226944 - 47227019 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0022732
miRBase ID: hsa-miR-3191-5p Mature miRNA Sequence: CUCUCUGGCCGUCUACCUUCCA Chromosome Location: Chr. 19 - 47226944 - 47227019 [-] on Build GRCh38 |