Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco Whirl-Pak™ Hydrated Poly-Sponge Write-On Bag

Nasco™ Whirl-Pak™ Hydrated Poly-Sponge Write-On Bag includes a sterile glove, intended for use with environmental monitoring

Supplier:  Nasco B01591

Catalog No. 14-209-181

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

  • Each sponge is packed in a sealed 18-oz. (532 mL) write-on bag
  • Hydrated with 10 mL of HiCap™ Neutralizing Broth
  • Terminally gamma processed to guarantee sterility
  • Two-year shelf-life with ambient storage
Specifications

Specifications

Write-On
Not Autoclavable
532 mL
Clear, Blue
Polyethylene Bag, Polyurethane Sponge, HiCap™ Neutralizing Broth, Sterile Glove
Sterile
10 mL of HiCap™ Neutralizing Broth
20 Sponges/Pouch, 5 Pouches/Box
2.50 mil
23 cm
11 cm
Ambient (36° to 86°F/ 2° to 30°C)
18 oz.
Food Grade
Environmental Sampling
Polyethylene, Polyurethane
0.064 mm
100/Pk.
Terminally Gamma Processed
9 in.
4.5 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit