Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Reeve Angel 230 Creped Qualitative Circles
Provide reliable performance for non-critical industrial laboratory and student lab applications
$15.50 - $297.00
Specifications
Format | Disc |
---|---|
Material | Cellulose |
Particle Retention | 25 to 30 μm |
Grade | 230 |
Porosity | Coarse |
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
09-805-17D
|
Cytiva
5230330 |
33 cm |
Pack of 50 for $115.00
|
|
|||||
WTI-230-25
|
Cytiva
5230250 |
25 cm |
Pack of 50 for $70.00
|
|
|||||
WTI-230-11
|
Cytiva
5230110 |
11 cm |
Pack of 50 for $18.50
|
|
|||||
WTI-230-12-5
|
Cytiva
5230125 |
12.5 cm |
Pack of 50 for $21.25
|
|
|||||
WTI-230-24
|
Cytiva
5230240 |
24 cm |
Pack of 50 for $71.25
|
|
|||||
09-803A
|
Cytiva
5230090 |
9 cm |
Pack of 50 for $15.50
|
|
|||||
09-803D
|
Cytiva
5230150 |
15 cm |
Pack of 50 for $29.25
|
|
|||||
09-805-17B
|
Cytiva
5230185 |
18.5 cm |
Pack of 50 for $41.25
|
|
|||||
05-713-507
|
Cytiva
5230400 |
40 cm |
Pack of 50 for $195.00
|
|
|||||
05-713-508
|
Cytiva
5230500 |
50 cm |
Pack of 50 for $297.00
|
|
|||||
Description
Retain coarse particles.
- Made from cellulose
- Creped surface for fast filtration of coarse particles
- Particle retention: 25 to 30μm
- Thickness: 0.40mm
Should not be used for kjeldahl nitrogen analysis
Specifications
Disc | |
25 to 30 μm | |
Coarse |
Cellulose | |
230 | |
0.4 mm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title