Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Water Pollution Kit
For detecting water pollution in aquatic environments.
$559.00
Specifications
For Use With (Application) | For detecting water pollution in aquatic environments |
---|---|
Grade | 5+ |
No. of Tests | 50 |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
S04351
|
LaMotte™
591703 |
Water Pollution 1 Kit |
Each of 1 for $559.00
|
|
|||||
Description
Water Pollution 1 Kit
- Examines water quality in suspected or known polluted aquatic environments
- Contains individual test modules for 11 key test factors: dissolved oxygen, chlorine, pH, nitrate-nitrogen, phosphate, ammonia-nitrogen and temperature
- With complete instructions and reagents to perform 50 tests for each factor
Titration Tests:
Test Factor / Range / Individual Kit Code
Dissolved Oxygen, 0-10.0 ppm (50 tests), 5860-01
Colorimetric Tests:
Test Factor / Range / Individual Kit Code
Chlorine, 0.2-3.0 ppm (50 tests), 3308-01
pH, 3-10.5 (100 tests), 5858-01
Nitrate-Nitrogen, 0.2-1.0 ppm (40 tests), 3119-01
Phosphate, 0.2-1.0 ppm (50 tests), 3119-01
Ammonia-Nitrogen, 1.0-4.0 ppm (50 tests), 5864-01
Temperature, -5°C–45°C, 1066
Order Info
Grades 5+.
Specifications
For detecting water pollution in aquatic environments | |
5+ | |
50 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title