Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Watch Glass, Economy
SDPSCI_ED

Watch Glasses featuring polished edges for safe use in the laboratory.

Supplier:  United Scientific™ WGL050

 View more versions of this product

Catalog No. S00834


Explore available promotions
Add to Cart
This item is not returnable. View return policy

Description

Description

  • Made from clear flint glass (1.8 to 2.0 mm thick)
  • Fine ground and polished edges
Specifications

Specifications

Autoclavable
Reusable
Round
Non-treated
2 in.
Round
Flint Glass
Non-sterile
Watch Glass
50 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.