Learn More
VistaLab™ Technologies Nozzle Filter Replacement Kits
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Nozzle filter kit for Macro model 0.2-10μL
Supplier: VistaLab™ Technologies 90606009
Description
- Replacement nozzle contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into the pipet
- If filter becomes wet, pipet will not aspirate fluid until a new filter or replacement nozzle is installed
Specifications
Non-sterile | |
0.2 to 10 mL Macro Pipette | |
5 in. | |
1 in. |
10 mL | |
5/Bag | |
3 in. |