Learn More
Gibco™ Virus Transport Medium
A ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B.
$4675.00
Specifications
Concentration | 1 X |
---|---|
Form | Liquid |
Product Type | Virus Transport Medium |
Serum Level | Standard Serum Supplementation |
Sterility | Sterile-filtered |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
A48750BA
![]() |
Gibco™
A48750BA |
1 L |
Each for $4,675.00
|
|
A48750DK
![]() |
Gibco™
A48750DK |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Kit for transporting samples suspected to contain virus
Gibco Viral Transport Medium is suitable for the transport of samples suspected to contain virus. It is a ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B.Specifications
1 X | |
Virus Transport Medium | |
Sterile-filtered | |
Dry Ice |
Liquid | |
Standard Serum Supplementation | |
No Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate |
Safety and Handling
ShelfLife : 12 months from date of manufacture
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.