Learn More
Gibco™ Virus Transport Medium
A ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B.
Supplier: Gibco™ A48750DK
Description
Kit for transporting samples suspected to contain virus
Gibco Viral Transport Medium is suitable for the transport of samples suspected to contain virus. It is a ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B.Specifications
Animal Origin | |
Liquid | |
Standard Serum Supplementation | |
No Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate |
1 X | |
Virus Transport Medium | |
Sterile-filtered | |
Dry Ice |