Learn More
Remco™ Vikan™ UST Long Handle Scrubbing Brush
Remco™ Vikan™ UST Long Handle Scrubbing Brush has the efficiency of a full-bristled brush without sacrificing cleanability and hygienic design.
Supplier: Remco™ 41962
Description
- Stiff bristles easily scrub away thought debris
- Ergonomically friendly handle makes the work easier
- Effective when cleaning machines, buckets, containers, and tubs in high risk areas
- The angled and circular arrangement of the Bristle Security Units enables efficient scrubbing and washing process
- Color-coded
Specifications
Scrub Brush | |
Stiff | |
Polypropylene | |
FDA Compliant | |
15.5 in. | |
Long |
Polyester | |
Yes | |
Non-sterile | |
Green | |
1 Ea. |