Learn More
Remco™ Vikan™ Extra Stiff Detail Brush
Remco™ Vikan™ Extra Stiff Detail Brush is designed with a smooth, ergonomic handle for cleaning in and around the gaskets and joints of machinery.
Supplier: Remco™ 44015
Description
- Small and compact design
- Suitable for cleaning in and around gaskets, rubber strips, and joints on machinery
- Can also be used as a grout brush and for cleaning equipment link conveyor belts
- Color-coded
Specifications
Cleaning Brush | |
Extra Stiff | |
Polypropylene | |
FDA Compliant | |
7.9 in. | |
Small |
Polyester | |
Yes | |
Non-sterile | |
White | |
1 Ea. |