Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Remco™ Vikan™ Dustpan
Makes for easier pick up of the debris.
$26.75 - $27.50
Catalog Number | Mfr. No. | Color Code | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color Code | Price | Quantity & Availability | |||||
19-830-056
|
Remco™
56603 |
Blue |
Each for $27.50
|
|
|||||
19-830-057
|
Remco™
56604 |
Red |
Each for $27.50
|
|
|||||
19-830-058
|
Remco™
56606 |
Yellow |
Each for $26.75
|
|
|||||
Description
- Features sharp edge so food particles are efficiently brushed into the pan
- Robust neck joint to withstand strong downward pressure
- Working Temperature: −30° to 80°C (−22° to 176°F)
Specifications
Dustpan | |
Yes | |
Polypropylene |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title