Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Remco™ Vikan™ Dustpan
Makes for easier pick up of the debris.
Supplier: Remco™ 56604
Description
- Features sharp edge so food particles are efficiently brushed into the pan
- Robust neck joint to withstand strong downward pressure
- Working Temperature: −30° to 80°C (−22° to 176°F)
Specifications
Red | |
Dustpan | |
Yes | |
Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction