Learn More
DWK Life Sciences Kimble™ ValueWare™ Volumetric Flasks, Class B, with Snap Cap
Calibrated “To Contain.”
Supplier: DWK Life Sciences 5801050
Description
- Kimax™ KG-33 glass
- Graduation ring blasted on neck, tooled for included polyethylene snap cap
- Enlarged top of the cap protects neck if flask is tipped over
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E288, Class B, unserialized requirements.
Specifications
Calibrated To Contain | |
50 mL | |
3 |
Polyethylene | |
Glass |