Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Vacuum Filtering Flasks
SDPSCI_ED

For use with 47 mm Vacuum Filtration Sets (FHFT47, FHPT47, and FHSS47)

$56.00 - $246.00

Specifications

Material Borosilicate Glass
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Rimmed
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S39253
SDPSCI_ED
View Documents
United Scientific Supplies
FHFL1000
1000 mL
Each for $56.00
Add to Cart
 
S39254
SDPSCI_ED
View Documents
United Scientific Supplies
FHFL4000
4000 mL
Each for $246.00
Add to Cart
 
Description

Description

  • Available in various capacities
  • Made from borosilicate glass
  • Suitable for use with most 47 mm Vacuum Filtration Sets
  • Compatible with United Scientific items: FHFT47, FHPT47, and FHSS47
Specifications

Specifications

Borosilicate Glass
Reusable
Rimmed
B
Stopper
Autoclavable
No
Non-sterile
Flat
None
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit