Learn More
Innovating Science™ Understanding Blood Type Interactions Through Simulated Blood Typing Kit

Use for deeper understanding of the importance of antigen/antibody interactions.
$28.75 - $56.00
Specifications
Color | White |
---|---|
Grade | 7 to 14 |
Age | 13+ years |
Includes | Teacher's manual, student guide, 40 blood typing trays, 4 simulated blood samples, donor no. 1, donor no. 2, donor no. 3, patient, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 1 bottle simulated anti-rh serum |
Quantity | 1 |
Includes:
- Four simulated blood samples
- One bottle each anti-A, anti-B and anti-Rh serum
- 40 blood typing trays
Description
Use simulated blood to determine the ABO/Rh blood type of four individuals, one in need of transfusion. The result determines which of three potential donors would provide the best match for the patient in need.
- Provides simulation of blood typing procedure
- No toothpicks, stirring or waiting for results
- Hands-on activities
- Kit contains materials for 10 groups
Kit uses simulated blood only.
Specifications
White | |
13+ years | |
1 |
7 to 14 | |
Teacher's manual, student guide, 40 blood typing trays, 4 simulated blood samples, donor no. 1, donor no. 2, donor no. 3, patient, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 1 bottle simulated anti-rh serum | |
10 Lab Groups |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.