Learn More
Innovating Science™ Understanding Blood Type Interactions Through Simulated Blood Typing Kit
Use for deeper understanding of the importance of antigen/antibody interactions.
Supplier: Innovating Science™ IS3103
Includes:
- Four simulated blood samples
- One bottle each anti-A, anti-B and anti-Rh serum
- 40 blood typing trays
Description
Use simulated blood to determine the ABO/Rh blood type of four individuals, one in need of transfusion. The result determines which of three potential donors would provide the best match for the patient in need.
- Provides simulation of blood typing procedure
- No toothpicks, stirring or waiting for results
- Hands-on activities
- Kit contains materials for 10 groups
Kit uses simulated blood only.
Specifications
49 CFR 173.4 | |
White | |
13+ years | |
Teacher's manual, student guide, 40 blood typing trays, 4 simulated blood samples, donor no. 1, donor no. 2, donor no. 3, patient, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 1 bottle simulated anti-rh serum | |
10 Lab Groups |
Simulated Blood Typing Kit | |
7 to 14 | |
For deeper understanding of the importance of antigen/antibody interactions | |
1 |