Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Type II Collagenase from Clostridium histolyticum
Used for heart, bone, muscle, thyroid, cartilage, and liver cells
Supplier: MP Biomedicals™ 02100502.5
Description
- Source: Clostridium histolyticum
Specifications
Brown, lyophilized powder | |
Collagenase | |
500 mg | |
Clostridium histolyticum |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction