Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Type II Collagenase from Clostridium histolyticum
Used for heart, bone, muscle, thyroid, cartilage, and liver cells
$75.90 - $2350.00
Specifications
Form | Brown lyophilized powder |
---|---|
Product Type | Collagenase |
Source | Clostridium histolyticum |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP21005023
|
MP Biomedicals™
02100502.3 |
25 mg |
Each for $75.90
|
|
MP21005021
|
MP Biomedicals™
02100502.1 |
100 mg |
Each for $132.30
|
|
MP21005025
|
MP Biomedicals™
02100502.5 |
500 mg |
Each for $387.50
|
|
MP210050205
|
MP Biomedicals™
0210050205 |
5 g |
Each for $2,350.00
|
|
Description
- Source: Clostridium histolyticum
Specifications
Brown lyophilized powder | |
Collagenase | |
Clostridium histolyticum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title