Learn More
Cardinal Health™ Two Color Medical Sterilization Wrap
Supports simultaneous double-wrapping to improve productivity
Supplier: Cardinal Health™ CH6G0045
Description
- SMS polypropylene material
- Two layers SMS (blue and green) bonded along opposing sides
- Colors used to differentiate, identify and inspect trays
- Cross-stitch pattern
- Clear, visual feedback of breaches that may occur during handling, storage and/or sterilization
Specifications
CH600 | |
Blue, Green | |
48/Cs. |
Two-color Sterilization Wrap | |
SMS Fabric, Polypropylene | |
45 x 45 in. |