Learn More
Tungsten wire, chromium plated, 1.8mm (0.07in) min dia, W 96.5+%, Cr 99.888% (metals basis), Thermo Scientific Chemicals
Evaporation source, contacts and coatings
$40.65 - $99.00
Specifications
Chemical Name or Material | Tungsten wire, chromium plated |
---|---|
Assay Percent Notes | W ≥96.5%, Cr 99.888% (metals basis) |
MDL Number | MFCD00011461 |
Form | Wire |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA41040BS
|
Thermo Scientific Chemicals
041040BS |
10 cm |
Each
for $40.65
Save 14% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA410408G
|
Thermo Scientific Chemicals
0410408G |
5 x 10 cm |
Each for $99.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Evaporation source, contacts and coatings
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tungsten wire, chromium plated | |
W ≥96.5%, Cr 99.888% (metals basis) | |
MFCD00011461 | |
Wire |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.