Learn More
Tungsten wire, chromium plated, 1.8mm (0.07in) min dia, W 96.5+%, Cr 99.888% (metals basis), Thermo Scientific Chemicals
Evaporation source, contacts and coatings
Supplier: Thermo Scientific Chemicals 0410408G
Description
Evaporation source, contacts and coatings
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Tungsten wire, chromium plated | |
W ≥96.5%, Cr 99.888% (metals basis) | |
5 x 10 cm |
≤1.8mm (0.07 in.) dia., thick: 1.3mm (0.05 in.), Cr layer: ≤0.01 in. thick | |
MFCD00011461 | |
Wire |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only