Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tufpak Secura'T™ Autoclave Bags
Clear film, printed bag with temperature indicator patch.
Supplier: Tufpak 11121924 PIO
Description
- Made from SUPROP™ Polypropylene film, specifically engineered for processing biohazardous materials and waste products in an autoclave prior to disposal
- Recommended for maximum temperature exposure to 140°C (285°F)
- Twist ties are included to allow proper closure for normal bag expansion and steam venting during sterilization cycle
- Bag exceeds strength requirements of 165g dart drop standard as performed under ASTM D1709-04
Specifications
Clear | |
Polypropylene Film | |
19 x 24 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction