Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tufpak Secura'T™ Autoclave Bags
Clear film, printed bag with temperature indicator patch.
$304.00 - $1211.00
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
14-375-210
|
Tufpak
11120812 PIO |
8 x 12 |
Case of 200 for $304.00
|
|
|||||
14-375-211
|
Tufpak
11121924 PIO |
19 x 24 |
Case of 200 for $726.50
|
|
|||||
14-375-212
|
Tufpak
11122535 PIO |
25 x 35 |
Case of 200 for $1,211.00
|
|
|||||
Description
- Made from SUPROP™ Polypropylene film, specifically engineered for processing biohazardous materials and waste products in an autoclave prior to disposal
- Recommended for maximum temperature exposure to 140°C (285°F)
- Twist ties are included to allow proper closure for normal bag expansion and steam venting during sterilization cycle
- Bag exceeds strength requirements of 165g dart drop standard as performed under ASTM D1709-04
- Color: Clear
Specifications
Clear | |
Polypropylene Film |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title