Learn More
Thermo Scientific™ TSX ultra-low freezer package with racks and boxes PROMO
Includes one ULT freezer and shelves of sliding drawer racks and boxes
Supplier: Thermo Scientific™ TSX60086DRAKQ1
Description
Package includes:
- One Thermo Scientific TSX Ultra-Low temperature freezer
- 2 shelves of sliding drawer racks with 2-inch cryoboxes and 100-count cell dividers. (see product specifications for details)
Specifications
Ultra-Low Freezer, Upright | |
Upright | |
37.6 x 39.6 x 78 in. |
-50°C to -86°C | |
600 box capacity freezer, 12 sliding drawer racks, 300 2-inch boxes with 100-count cell dividers | |
208 - 230 V 60 Hz |